NMR sample tubes, 10 mm, PYREX®, CORNING®
Supplier: Corning
Jump to:
About this item
Made of PYREX® borosilicate glass.
These precision 10 mm diameter, 7 inch long PYREX® NMR tubes are compatible with ground joints, valves, and vacuum racks. They are designed for use on NMR spectrometers running from 400 - 600 MHz.
Outer Ø: 10,00 + 0,000 to 0,013 mm
Internal Ø: 9,07 ±0,013 mm
2 Options Available Below
Product Details & Documents
Made of PYREX® borosilicate glass.
These precision 10 mm diameter, 7 inch long PYREX® NMR tubes are compatible with ground joints, valves, and vacuum racks. They are designed for use on NMR spectrometers running from 400 - 600 MHz.
Outer Ø: 10,00 + 0,000 to 0,013 mm
Internal Ø: 9,07 ±0,013 mm