Measuring pipettes (Mohr), class A, reusable, PYREX®
Supplier: Corning
Certificates
About this item
These reusable PYREX® glass pipettes are colour-coded by size for easy identification and sorting.
- Calibrated 'to deliver' (TD) their total capacity without blow-out
- Top end of 5 to 25 ml sizes is constricted (Mohr type)
- Coloured graduations are enameled onto the glass
- Colour-coded by size for easy identification
- Each pipette is individually serialised and certified
Certifications: Calibrated to Class A tolerances in accordance with ASTM E-542, ATSM E-694, and ASTM E-1293 specifications.
Delivery information: Supplied with a certificate of identification and capacity, traceable to NIST standards.