Barrett distilling apparatus with stopcock and 24/40 standard taper joints, PYREX®
CORN3622-20
: Corning
- Cone size:24/40 NS
- Jacket length:228 mm
- Socket size:24/40 NS
- Pk:1
This PYREX® distilling apparatus is designed for determining water content in petroleum or bituminous products.
- With stopcock (glass plug) and 24/40 [ST] joints
- Very good temperature resistance
- Very good chemical resistance
It is graduated upwards from the stopcock to 3 ml, in 0,2 ml divisions, and from 3 ml to 20 ml in 0,5 ml divisions.
The apparatus can also be used as Dean Stark distillation apparatus specified in British Standard 756, but utilises a common U.S. ground joint. It is also an optional setup for ASTM D-95.