Order Entry
Singapore
ContactUsLinkComponent
Dropping bottles, with bulb and pipette stopper, PYREX®
Dropping bottles, with bulb and pipette stopper, PYREX®
  CORN1340-125
 :  Corning
undefined
Dropping bottles, with bulb and pipette stopper, PYREX®
  CORN1340-125
 :  Corning

 

  • Ext.Ø:
    55 mm
  • Capacity:
    125 ml
  • Height:
    150 mm
  • Seq:
    01
  • Pk:
    12

 

 

Narrow mouth droppings bottles, made of PYREX® borosilicate glass.

  • Reusable bottle
  • Pipette with suction bulb
  • Hollow glass stopper style

Recommended for use where preventing contamination by solubility or pH change is important.