Media bottles, wide mouth, graduated, with plug seal screw cap, PYREX®
Supplier: Corning
Certificates
About this item
Bottles, made of PYREX® borosilicate glass, with orange PP screw cap.
- Graduated
- Very good chemical and temperature resistance
- PP screw cap design permits faster opening and closing
- Comfortable and ergonomic cap handling
- Indented gripping zones along the sides of bottle allow easier and safer handling
Provide easier handling and more efficient storage of media, biologicals, and other aqueous solutions. Slim Line media storage bottles can be used for storage as well as mixing. Widely spaced knurls provide comfortable and ergonomic cap handling. Narrow bottle diameter allows more efficient storage on shelves, refrigerators, autoclaves.