Order Entry
Singapore
ContactUsLinkComponent
 
Chloroform ≥99.0% stabilised BP, USP, NF for UV spectrophotometry, for IR spectrophotometry, for HPLC
Catalog # APPC361252.1612
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Chloroform ≥99.0% stabilised BP, USP, NF for UV spectrophotometry, for IR spectrophotometry, for HPLC
Catalog # APPC361252.1612
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Appearance:
    Clear, colourless liquid
  • Solubility:
    Soluble in water 8 g/l at 20 °C
  • Vapour pressure (20 °C):
    210 hPa
  • Viscosity (20 °C):
    0,56 mPa.s
  • Dipole moment (20° C):
    1,01 Debye
  • Dielectric constant (20 °C):
    4,8
  • Heat of evapouration (61 °C ):
    263 kJ/kg
  • Saturation concentration (20 °C):
    1027 g/m3
  • Minimum assay (G.C.):
    99,0%
  • APHA colour:
    10
  • Acidity:
    0,00015 mEq/g
  • Alkalinity :
    0,0002 mEq/g
  • Non-volatile matter  :
    0,0003%
  • Ethanol (G.C.):
    0,5 - 0,8%
  • Water (H₂O):
    0,01%
  • Suitability for IR spectrometry:
    Passes test
  • Fluorescence at 254 nm (as quinine):
    1 ppb
  • Fluorescence at 365 nm (as quinine):
    1 ppb
  • UV Spectrum (1cm cell; Ref.: water)::
    0
  • Transmittance at 244 (Cut off) nm:
    ≥10%
  • Transmittance at 245 nm:
    ≥20%
  • Transmittance at 250 nm:
    ≥50%
  • Transmittance at 257 nm:
    ≥80%
  • Transmittance at 260 nm:
    ≥85%
  • Transmittance at 270 nm:
    ≥90%
  • Transmittance at 280-400 nm:
    ≥98%
  • Data of interest in HPLC::
    0
  • P' + 0.25 E:
    5,6
  • Rohrschneider Polarity:
    4,1
  • Eluotropic value e° (Al₂O₃):
    0,4
  • Sol. H₂O in solv. at 20 °C:
    0,072

Specification Test Results

About this item

 :
 :119.38 g/mol
Boiling Pt:61 °C
Melting Pt:−63 °C
Melting Pt:–63 °C
Density:1,476…1.486
UN:1888
ADR:6.1,III
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826