

pH Indicator solution universal pH 3-10, Fluka™
HONC31282-100ML
:
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
- Shade of color according to scale:complying
:
:0,89 g/cm³ (20 °C)
:19 °C
:Ambient
:1170
:3,II
:GHS07
:GHS02
:MFCD00148741