Human IgM Isotype Control
Catalog # ICNA0855916
Supplier: MP Biomedicals
Specifications
- Antibody type:Control
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Human
- Isotype:IgM
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Concentration:Total Protein: 5.3 mg/mL; Volume: 19.4 mL
- Shipping temperature:RT
- Purification:Human IgM is purified from human myeloma serum using multi-step procedures which may include salt fractionation, gel filtration, ion-exchange chromatography and immunoabsorption. The product is dialyzed into 0.01M sodium phosphate, 0.07M sodium chloride, pH 7.3, filtered through a 0.22 μm filter and vialed. No preservative has been added.
- Size:5 mg
- Pk:5 mg
Specifications
About this item
Product is human IgM and buffer salts.The product is tested for purity at a minimum of 20 mg/mL using immunoelectrophoresis. The product is human IgM; no trace of IgG, IgA or non-immunoglobulin serum proteins is detected. The total protein is determined using the Biuret procedure, with bovine albumin as the standard. Each vial is overfilled by approximately 10 percent so that a minimum of 5 mg of human IgM is recovered.