Specifications
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- ELISA:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Rabbit
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Concentration:Antibody Concentration: 0.6 mg/mL
- Shipping temperature:RT
- Purification:Purified by gel filtration. The product is dialyzed into 0.03M TRIS, 0.14M sodium chloride, pH 8.0, with 1% bovine albumin, adjusted to standard titer, filtered through a 0.22 µm filter, and vialed
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Product is alkaline phosphatase (AKP)-conjugated goat affinity purified antibody to rabbit IgG (whole molecule) and buffer salts. To ensure that the product titer falls within the required range, antibody titer is standardized by microtiter plate ELISA with rabbit IgG. The antibody is tested for purity at 10 mg/mL by immunoelectrophoresis. The antibody is goat IgG; no trace of albumin is detected.
Antibody and highly purified calf-intestine alkaline phosphatase are conjugated under defined conditions to obtain optimally labeled product. Conjugated protein is purified by gel filtration. The product is dialyzed into 0.03M TRIS, 0.14M sodium chloride, pH 8.0, with 1% bovine albumin, adjusted to standard titer, filtered through a 0.22 µm filter, and vialed
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AKP
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit