Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))
ICNA0855391
- Antibody type:Secondary
- Antigen symbol:IgG
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Guinea pig
- Form:Lyophilized
- Concentration:Total Protein: 15.9 mg/mL Titer: 1:176,000
- Shipping temperature:RT
- Purification:Antibody and highly purified HRP (Rz>3.0) are conjugated under defined conditions to obtain optimally labeled product. Conjugated protein is purified by salt fractionation. The product is dialyzed into 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, adjusted to standard titer, filtered through a 0.22 μm filter, vialed and lyophilized. No preservative has been added.
- Size:2 ml
- Pk:2 mL
Product is the lyophilized powder of horseradish peroxidase (HRP)-conjugated goat IgG fraction to guinea pig IgG (whole molecule) and buffer salts. The total protein is measured using the Biuret procedure with bovine albumin as the standard. To ensure that the product titer falls within the required range, antibody titer is standardized by microtiter plate ELISA with guinea pig IgG. The product is tested for purity and specificity at final concentration by immunoelectrophoresis. The antibody is predominantly goat IgG; no trace of albumin is detected.
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Guinea pig