Order Entry
Singapore
ContactUsLinkComponent
 
Anti-IgA alpha Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  ICNA0855077
Anti-IgA alpha Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  ICNA0855077

 

  • Antibody type:
    Secondary
  • Antigen name:
    alpha-chain specific)-FITC 
  • Antigen symbol:
    IgA alpha
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgA
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Lyophilized powder
  • Storage buffer:
    pH 7.3
  • Concentration:
    Total Protein: 47.8 mg/mL; Antibody Protein: 6 mg/mL
  • Shipping temperature:
    RT
  • Purification:
    Antibody and FITC are conjugated under defined conditions to obtain the specified molar ratio (3-6 mol/mol). Conjugated protein is purified by gel filtration. The product is dialyzed into 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, with 0.05% sodium azide, adjusted to standard titer, filtered through a 0.22 µm filter, vialed, and lyophilized
  • Size:
    2 ml
  • Pk:
    2 mL

 

 

IgA antibody is a secretory antibody and is present abundantly in mucous linings of gastrointestinal, respiratory and genitourinary tracts, tears and saliva. Thus, secretory IgA contributes to the humoral defense mechanism against the pathogens on mucosal surfaces.

Type: Secondary
Antigen: IgA alpha
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgA
Reactivity: Human