Capillary columns, SolGel-WAX™
These columns with polyethylene glycol (PEG) in a Sol-Gel matrix are recommended for highly active compounds.
- Less susceptible to damage by oxygen than conventional wax phases
- Polar phase
- Low bleed and inert
Suitable replacement for: DB-Wax, Rtx-Wax, Stabilwax, HP20M, HP-Wax, HP-INNOWax, Supelcowax-10, AT-Wax, Nukol, CP Wax 52CB, VB-WAX, ZB-WAX.
Maximum temperature: 30 °C to 260/280 °C
Polyethylene glycol
Max. temperature: 0,1 - 1,0 µm film thickness 20 °C to 260/280 °C; >1,0 µm film thickness 20 °C to 240/260 °C