Q-VD-OPh, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Specifications
- CAS No.:1135695-98-5
- Storage conditions:+4 °C
- MDL No.:MFCD20527311
- Protein synonyms:Q-Val-Asp-OPh
- Protein/peptide name:Q-VD-OPh
- Molecular weight:513,5 g/mol
- Molecular formula:C₂₆H₂₅F₂N₃O₆
- Purity:95%