AlbumiNZ™ Bovine serum albumin (BSA), MP Biomedicals
Catalog # ICNA0219989890
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:500 g
- CAS No.:9048-46-8
- Source:Serum
- Species:Bovine
- Seq:04
- Storage conditions:+4 °C
- Form:Powder
- Protein synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/peptide name:Bovine Serum Albumin
- Purity:>97%
- pH:6,5 - 7,5
- Grade:Immunoassay (EIA) grade
Specifications
About this item
AlbumiNZ™ Protease-reduced BSA is useful for immunoassays where proteases may degrade proteins or where peroxidase or alkaline phospahatase may hydrolyse substrates.
AlbumiNZ™ Protease-reduced BSA is chromatographically purified from New Zealand-sourced bovine plasma in an ISO quality system assuring complete traceability and consistent high quality. There is no detectable enzymatic activity.
Biochem/physiol actions: Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.