Human beta2-Microglobulin (from Urine), MP Biomedicals
Catalog # ICNA02153903.5
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:500 µG
- Source:Urine
- Species:Human
- Storage conditions:–20 °C
- Protein/peptide name:beta2-Microglobulin
- Purity:30 - 60 % (Total Protein)
- Molecular weight:11,800
Specifications
About this item
- Lyophilized
- Purity: 30 to 60% total protein
Storage temperature: -20 °C