Agar (from Red Alga Rhodophyceae), off-white powder, Bacteriological Grade
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:9002-18-0
:
MDL Number:MFCD00081288
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Salmonella:Negative
- Loss on Drying:≤20%
- Total Ash (dry weight basis):≤6.5%
- Acid Insoluble Ash:≤0.5%
- Foreign Organic Matter:≤1.0%
- Foreign Insoluble Matter:≤1.0%