Snap-on circuit breaker lockout
Catalog # BRDY65387
Supplier: Brady
Specifications
- Colour:Red
- W×D×H:8,9×54,6×21 mm
- Pk:1
Specifications
About this item
This snap-on breaker lockout device isolates and secures circuit breaker switches to protect employees against electrical accidents during equipment maintenance.
- Easy and quick to install, just snap in place and apply lock
- Device accepts lock shackles up to ⁹/₃₂" in diameter
It is designed for 120 V circuit breakers that have holes in the switch tongue.