Recombinant IL2 (from E. coli), MP Biomedicals
Catalog # ICNA0215389610
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:10 KU
- Protein/peptide type:Recombinant
- Species:E. coli
- Storage conditions:−20 °C
- Carrier-free:Y
- Protein synonyms:Interleukin-2
- Protein/peptide name:IL2
- Purity:>95%
- Molecular weight:17 kDa
- Colour:White
- Formulation:Lyophilised
- Grade:Ultrapure grade
Specifications
About this item
Interleukin-2 is an immunomodulatory factor produced by certain subsets of T lymphocytes. This lymphokine promotes long term growth of activated T cells and related cell types.
- Specific activity: ≥5×10⁶ µ/mg
- Endotoxin: ≤0,15 ng/µ
It plays a role in the activation and proliferation of NK cells, induces γ-interferon and B cell growth factor secretion, and modulates the expression of the IL-2 receptor.