Order Entry
Singapore
ContactUsLinkComponent
Bovine growth hormone (from Pituitary glands), MP Biomedicals
Bovine growth hormone (from Pituitary glands), MP Biomedicals
Catalog # ICNA02160074.1
Supplier:  MP Biomedicals
CAS Number:  
Bovine growth hormone (from Pituitary glands), MP Biomedicals
Catalog # ICNA02160074.1
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Source:
    Pituitary glands
  • Species:
    Bovine
  • Storage conditions:
    −20 °C
  • Form:
    Powder
  • Protein/peptide name:
    Growth Hormone
  • Molecular weight:
    24 kDa
  • Colour:
    White

Specifications

About this item

Bovine growth hormone is a peptide hormone, it is used in regulating metabolic processes.

Biological activity: In responsive cell lines, biological activity is observed at concentrations of 10 to 100 ng/ml.