Immunoblotting detection kits, Immu-Mark™ Silver Enhancing Kit, MP Biomedicals
Catalog # ICNA08678211
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:500 mL
- Description:Immu-Mark™ Silver Enhancing Kit
Specifications
About this item
Immu-Mark™ Silver Enhancing Kit is used for amplifying the signal from gold particles attached to proteins or antigens immobilised on membranes. By intensifying the gold probe signal, protein levels in the 1 picogram range are easily detectable.
- Easy to use
- Sensitive and rapid method
- Intense staining with minimal background
- Darkroom is not necessary for this procedure