Bovine casein (from Milk), MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Casein from bovine milk is a phosphoprotein. It make up approximately 80% of the total protein in bovine milk. It is proposed to be the main protective constituent in milk.
It is used in animal diet and as a blocking agent in immunochemistry. As a food source, casein supplies amino acids; carbohydrates; and two inorganic elements, calcium and phosphorus.
Specifications
- CAS No.:9000-71-9
- Source:Milk
- Species:Bovine
- Storage conditions:+15 to +30 °C
- Form:Powder
- Protein/peptide name:Casein
- Colour:Cream