Megathura crenulata Hemocyanin, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Hemocyanins are copper-containing oxygen transport proteins found in the hemolymph of many invertebrates.
- Water soluble
It is used to immunise animals to elicit antibodies to small molecules (haptens) by covalent conjugation. It is often used as a carrier protein due to its highly immunogenic properties and the large number of lysine residues available for modification. Soluble KLH is suitable for the preparation of immunogens for injection, or as a control protein for ELISA and dot blot immunoassay due to its high solubility in water and buffers.
Specifications
- CAS No.:9013-72-3
- Species:Megathura crenulata
- Storage conditions:+4 °C
- Protein/peptide name:Hemocyanin
- Colour:Grey
- Formulation:Lyophilised powder