Anti-Prostate Specific Antigen Mouse Monoclonal Antibody [clone: ER-PR8]
Catalog # ICNA08691881
Supplier: MP Biomedicals
Specifications
- Antibody type:Primary
- Antigen name:Prostate Specific Antigen
- Antigen symbol:PSA
- Clonality:Monoclonal
- Clone:ER-PR8
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Format:Ascites fluid
- Form:Liquid
- Storage temperature:–20 °C
- Tested applications:Immunoblot, Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections, Immunohistochemical Staining of Frozen Tissue Sections
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Immunogen was from human prostate. The antibody reacts with a 34 kDa protein. Immunohistochemical studies have shown that the presence of PSA is highly specific for the demonstration of the prostatic origin of a malignancy. Antibodies to PSA have been shown to be valuable reagents for the positive identification of primary and metastatic prostatic carcinoma.
- Working dilution: 1:10
- It reacts exclusively with prostatic epithelium and prostatic carcinoma cells
Type: Primary
Antigen: Prostate Specific Antigen
Clonality: Monoclonal
Clone: ER-PR8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human