Mitogen (from Pokeweed roots), MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Pokeweed mitogen is extracted from pokeweed roots. This protein has hemagglutinating, leukoagglutinating, and mitogenic properties. It is not blood group specific, but has affinity for N-acetyl-β-D-glucosamine oligomers.
- Soluble in phosphate buffered saline (1 mg/ml)
- Can be reconstitute with 0,01 M sodium phosphate, 0,15 M sodium chloride
- Potency: 0,03 to 0,3 μg/ml mitogenic activity
- Extinction Coefficient (E1%): E1%=18,5 (280 nm in water) (Lit.)
It shows agglutination when tested with human red blood cells.
Specifications
- Source:Pokeweed roots
- Storage conditions:+2 to +8 °C
- Protein/peptide name:Mitogen
- Formulation:Lyophilised powder