Order Entry
Singapore
ContactUsLinkComponent
Adapters, inlet, with cone
Adapters, inlet, with cone
  CGLACG-1014-01
 :  CHEMGLASS
undefined
Adapters, inlet, with cone
  CGLACG-1014-01
 :  CHEMGLASS
  • Cone size:
    24/40 NS
  • Pk:
    1
Adapters allow for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection has a 10 mm O.D. at the largest serration and is bent downward at a 90° angle to the cone.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes