Adapters, reduction
24/40 NS, 19/22 NS
Catalog # CGLACG-1000-44
Marketsource
Supplier: CHEMGLASS
Jump to:
Product Details & Documents
Used to connect dissimilar standard taper joint sizes. Reduction adapter from large cone at the base to small socket on the top.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
- Cone size:24/40 NS
- Socket size:19/22 NS
- Seq:01