Receiver adapters, vacuum take-off, bent
CGLACG-1050-01
- Cone size:24/40 NS
- Socket size:24/40 NS
- Environmentally Preferable:
- Pk:1
With standard ground joint cone and socket with sealed in drip tip. Adapter is bent at a 105° angle for use in vacuum distillation. Serrated hose connection has an O.D. of 10 mm at the largest serration for connection to vacuum source.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes