Order Entry
Singapore
Orders LinkContactUsLinkComponent
Fritted support assemblies

Fritted support assemblies

Complete support assembly, fritted, for 47 mm membranes
Catalog # CGLACG-1420-01
Marketsource
Supplier:   CHEMGLASS
Fritted support assemblies
Catalog # CGLACG-1420-01
Supplier:  CHEMGLASS
Marketsource
Fritted support assemblies
Fritted support assemblies

Complete support assembly, fritted, for 47 mm membranes

Price
$775.00
Log in for availability
1 * 1 items
Product Details & Documents

Glassware system for vacuum filtration using 47 mm membranes. Assembly is designed for use with vacuum manifolds or optional plastic coated filtering flask. Support base has a sealed-in coarse porosity fritted disc.

Delivery information: Complete assembly includes a 300 ml graduated funnel, aluminium clamp, support base and silicone stopper.

Specifications
  • Description:
    Complete support assembly, fritted, for 47 mm membranes
  • Seq:
    01