Order Entry
Singapore
ContactUsLinkComponent
Flasks, round bottom, glass, clear, standard ground joint, one neck
Flasks, round bottom, glass, clear, standard ground joint, one neck
Catalog # CGLACG-1506-17
Supplier:  CHEMGLASS
undefined
Flasks, round bottom, glass, clear, standard ground joint, one neck
Catalog # CGLACG-1506-17
Supplier:  CHEMGLASS
Specifications
  • Socket size:
    24/40 NS
  • Capacity:
    250 ml
  • Pk:
    1
Specifications
About this item
Heavy walled, single short neck round bottom flasks with standard taper outer joints.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes