Order Entry
Singapore
ContactUsLinkComponent
 
Bovine aprotinin (from Lung), MP Biomedicals
 
Bovine aprotinin (from Lung), MP Biomedicals

 

  • CAS No.:
    9087-70-1
  • Source:
    Lung
  • Species:
    Bovine
  • Storage conditions:
    +4 °C
  • Biological activity:
    ~6000 KIU/mg
  • Protein/peptide name:
    Aprotinin
  • Molecular weight:
    6,5 kDa
  • Molecular formula:
    C₂₈₄H₄₃₂N₈₄O₇₉S₇