Order Entry
Singapore
ContactUsLinkComponent
Chicken Trypsin inhibitor (from egg white), MP Biomedicals
Chicken Trypsin inhibitor (from egg white), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Chicken Trypsin inhibitor (from egg white), MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    9035-81-8
  • Source:
    Egg white
  • Species:
    Chicken
  • Storage conditions:
    +4 °C
  • Protein/peptide name:
    Trypsin inhibitor
  • Molecular weight:
    28,5 kDa

Specifications