Rabbit Actin (from Muscle), MP Biomedicals
ICNA08771012
:
- Pk:100 µG
- Source:Muscle
- Species:Rabbit
- Storage conditions:0 to +5 °C
- Protein/peptide name:Actin
- Purity:~98% (by SDS-PAGE)
- Molecular weight:43 kDa
- Formulation:Lyophilised powder
Actin is a purified protein from Rabbit muscle, for use as a high molecular weight marker or as a standard.
It may be used as an antigen for antibody development.