Büchner funnels
Marketsource
Supplier: CHEMGLASS
Jump to:
About this item
Büchner style flter funnels with a lower standard taper inner joint and serrated tubulation for connection to a vacuum source.
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
- Very good temperature resistance
For convenient filtration of solutions.
3 Options Available Below
Product Details & Documents
Büchner style flter funnels with a lower standard taper inner joint and serrated tubulation for connection to a vacuum source.
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
- Very good temperature resistance
For convenient filtration of solutions.
Product Family Options
Product Information
- Plate ØCapacityPorosityAvailabilityPrice
- CGLACG-1406-11Cone NS 24/40Specifications:MarketsourceMarketsource
- CGLACG-1406-25Cone NS 24/40Specifications:MarketsourceMarketsource
- CGLACG-1406-31Cone NS 24/40Specifications:MarketsourceMarketsource