Flasks, round bottom, glass, clear, standard ground joints, three necks
Made of borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Heavy wall three neck round bottom flasks with standard taper outer joints on the both center neck and 20° angled side necks.