Order Entry
Singapore
Orders LinkContactUsLinkComponent
Evaporating flasks
Marketsource
Supplier:   CHEMGLASS
Evaporating flasks
Supplier:  CHEMGLASS
Marketsource
Certificates
Evaporating flasks
Evaporating flasks
6 Options Available Below
Product Details & Documents
Made of borosilicate glass, coated.
  • Flasks with 24/40 joints have a square reniforcing bead
  • Plastic coated to prevent injury from broken glass, and also prevents spillage of hazardous materials
  • Very good chemical resistance
These single neck recovery flask can be used as an evaporating flask in rotary evaporators.
The flask have a heavy wall for vaccum applications and are balance to prevent run-out during rotation.
Product Family Options
Product Information
  • Capacity
    Socket size
    Availability
    Price
  • 10 ml
    14/20 NS
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $182.00
    $182.00
    1 * 1 items
       $182.00
    Specifications:
    Marketsource
    Marketsource
  • 25 ml
    14/20 NS
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $182.00
    $182.00
    1 * 1 items
       $182.00
    Specifications:
    Marketsource
    Marketsource
  • 50 ml
    14/22 NS
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $182.00
    $182.00
    1 * 1 items
       $182.00
    Specifications:
    Marketsource
    Marketsource
  • 50 ml
    24/40 NS
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $224.00
    $224.00
    1 * 1 items
       $224.00
    Specifications:
    Marketsource
    Marketsource
  • 100 ml
    24/40 NS
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $231.00
    $231.00
    1 * 1 items
       $231.00
    Specifications:
    Marketsource
    Marketsource
  • 200 ml
    24/40 NS
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $303.00
    $303.00
    1 * 1 items
       $303.00
    Specifications:
    Marketsource
    Marketsource