Adapters, glass, flushing, Airfree, Schlenk type
Catalog # CGLAAF-0509-11
Supplier: CHEMGLASS
Specifications
- Cone size:24/40 NS
- Type:with 4 mm glass stopcock
- Pk:1
Specifications
About this item
Adapter allows vessels to be flushed with inert gas while sampling directly through the bore of the T-Bore stopcock. With a 4 mm bore high vacuum glass stopcock with stainless steel constant pressure retention device.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Both the glass and PTFE stopcock have 9 mm OD medium wall arms for use with septum stopper.