Order Entry
Singapore
ContactUsLinkComponent
 
Human native transferrin (from plasma), MP Biomedicals
  ICNA08823431
 :  
Human native transferrin (from plasma), MP Biomedicals
  ICNA08823431
 :  

 

  • Pk:
    1 g
  • CAS No.:
    11096-37-0
  • Protein/peptide type:
    Native
  • Source:
    Plasma
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Protein/peptide name:
    Transferrin
  • Purity:
    99% (by SDS-PAGE)
  • Concentration:
    100% (by Lowry)
  • Formulation:
    Lyophilised powder
  • pH:
    6,8

 

 

Transferrin is a glycoprotein with homologous N-terminal and C-terminal iron-binding domains. It is ideal for use as a growth factor for primary, continuous, and transformed cell cultures.

  • Soluble in 50 mg/ml purified water
  • No IgG's were detected
  • Endotoxin level: <1 EU/mg by LAL