DNA extraction kits, MPure™ Plant DNA extraction kit, MP Biomedicals
Catalog # ICNA117022150
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:1 KIT
- Application:
- Method / Format:Magnetic beads
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:Plant samples
- Target molecule:DNA
- Description:MPure™ Plant DNA Extraction Kit, 48 tests
- Environmentally Preferable:
- Description:MPure™ Plant DNA Extraction Kit, 48 tests
Specifications
About this item
MPure™ Plant DNA Extraction Kit is used with the MPure-12™ Automated Nucleic Acid Purification System for extraction of genomic DNA from plant (leaf, seeds and spores) and fungal tissues. Up to 100 mg of tissue can be used for purification.