Order Entry
Singapore
ContactUsLinkComponent
Bovine IgG Isotype Control
Bovine IgG Isotype Control
Catalog # ICNA08641401
Supplier:  MP Biomedicals
Bovine IgG Isotype Control
Catalog # ICNA08641401
Supplier:  MP Biomedicals

Specifications

  • Antibody type:
    Control
  • Antigen symbol:
    IgG
  • Host:
    Bovine
  • Isotype:
    IgG
  • Reactivity:
  • Seq:
    01
  • Environmentally Preferable:
  • Storage temperature:
    2...8 °C
  • Pk:
    100 mg

Specifications

About this item

It is useful for structural and biological studies, as a reference standard, or as starting material for immunogen preparation. It is prepared using salt fractionation or by chromatographic techniques.

  • Immunoassay grade
  • Store at 2 to 8 °C