Order Entry
Singapore
ContactUsLinkComponent
 
AlbumiNZ™ Bovine serum albumin (BSA), MP Biomedicals
 
undefined
AlbumiNZ™ Bovine serum albumin (BSA), MP Biomedicals

 

  • CAS No.:
    9048-46-8
  • Source:
    Serum
  • Species:
    Bovine
  • Storage conditions:
    +2 to +8 °C
  • Form:
    Powder
  • Protein synonyms:
    BSA|Fraction V|Cohn Fraction V
  • Protein/peptide name:
    Bovine Serum Albumin
  • Purity:
    >97%
  • pH:
    6,5 - 7,5
  • Grade:
    Microbiological grade