ImmunO™ Bovine;Cat;Dog;Guinea pig;Human;Mouse;Rat;Sheep fibrinogen (from plasma), MP Biomedicals
Fibrinogen is a blood protein that is involved in clotting and is converted to fibrin by thrombin. Fibrinogen has an approximate molecular weight of 340 kDa. It consists of three non-identical pairs of disulfidebonded chains. The α-chain has an approximate molecular weight of 63,5 kDa, the β-chain 56 kDa, and the γ-chain 47 kDa. At the amino termini, the three chains are connected in a dimeric disulfide knot (DSK). A second DSK occurs later in the molecule. Fibrinogen has approximately 4% carbohydrate content.
- CAS No.:9001-32-5
- Source:Plasma
- Species:SheepHumanBovineGuinea pigDogRatMouseCat
- Storage conditions:Store product at <-10 °C
- Gene ID:522039, 510522, 280792
- Protein synonyms:Factor 1
- Protein/peptide name:Fibrinogen
- Tested applications:Substrate for clotting assays