Order Entry
Singapore
ContactUsLinkComponent
Anti-Bovine Desmoplakin I/II Mouse Monoclonal Antibody [clone: Dp 2.15]
Anti-Bovine Desmoplakin I/II Mouse Monoclonal Antibody [clone: Dp 2.15]
Catalog # ICNA08695421
Supplier:  MP Biomedicals
Anti-Bovine Desmoplakin I/II Mouse Monoclonal Antibody [clone: Dp 2.15]
Catalog # ICNA08695421
Supplier:  MP Biomedicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Desmoplakin I/II
  • Clonality:
    Monoclonal
  • Clone:
    Dp 2.15
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Chicken,
    Human,
    Bovine,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilised powder
  • Storage temperature:
    –20 °C
  • Tested applications:
    Immunohistochemical Staining of Frozen Tissue Sections
    Immunoblot
    Indirect Immunofluorescence
  • Purification:
    Affinity purified
  • Size:
    50 μg
  • Pk:
    50 µG

Specifications

About this item

This antibody stains desmosomes and is an excellent histological marker for epithelial tissues and epithelium-derived tumors including all primary and metastic carcinomas. In combination with cytokeratin antibodies, they may aid in the identification of epithelial origin tumors and metastases. It has been shown to hold value in meningioma identification.

  • Can be reconstitute with 1,0 ml of distilled water
  • Dilution range: 1:10 with PBS for Immunohistochemistry
  • Tumors specifically detected: Primary and metastatic Carcinoma and Meningioma

Type: Primary
Antigen: Desmoplakin I/II
Clonality: Monoclonal
Clone: Dp 2.15
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Chicken, Human, Mouse, Rat