Order Entry
Singapore
ContactUsLinkComponent
Anti-Synaptophysin IgG1 Mouse Monoclonal Antibody [clone: SY38]
Anti-Synaptophysin IgG1 Mouse Monoclonal Antibody [clone: SY38]
Catalog # ICNA08697301
Supplier:  MP Biomedicals
Anti-Synaptophysin IgG1 Mouse Monoclonal Antibody [clone: SY38]
Catalog # ICNA08697301
Supplier:  MP Biomedicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Synaptophysin
  • Antigen symbol:
    SYP
  • Clonality:
    Monoclonal
  • Clone:
    SY38
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Bovine,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilised
  • Molecular weight:
    38 kDa
  • Storage temperature:
    +2 °C to +8 °C
  • Immunogen:
    Synaptophysin from presynaptic vesicles of bovine brain
  • Tested applications:
    Immunoblot, Fluorescence-Activated Cell Sorting, Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections, Immunohistochemical Staining of Frozen Tissue Sections
  • Purification:
    Affinity purified
  • Size:
    50 μg
  • Pk:
    50 µG

Specifications

About this item

This antibody identifies synaptophysin, a 38 kDa membrane glycoprotein. It may be a valuable aid in the examination of various neuropathological disorders, especially those associated with altered synapse patterns and defects of neurotransmitter sections. It may also possess potential for studying diseases such as Alzheimer's disease, alcoholic fetoembryopathy, and senile dementia.

  • Specificity: It is an excellent marker for several neuroendocrine, neuronal and adrenal tumors, and reacts with synaptophysin, a transmembrane glycoprotein of presynaptic vesicles
  • Dilution range: 1:50 for Immunohistochemical application
  • It can reconstitute with 1,0 ml of distilled water

The antibody is in PBS buffer, pH 7,4, with 0,5% BSA, and 0,09% sodium azide.

Type: Primary
Antigen: Synaptophysin
Clonality: Monoclonal
Clone: SY38
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Rat, Rat PC-12 cell line