Anti-Chondroitin-4-Sulphate Mouse Monoclonal Antibody [clone: BE-123]
Catalog # ICNA08636511
Supplier: MP Biomedicals
Specifications
- Antibody type:Primary
- Antigen name:Chondroitin-4-Sulfate
- Clonality:Monoclonal
- Clone:BE-124
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Format:Ascites fluid
- Form:Liquid
- Storage temperature:–20 °C
- Concentration:Conc/Titer: 1:50-1:6, 400(Lit.)
- Shipping temperature:Dry Ice
- Immunogen:Mouse proteoglycan
- Tested applications:Radio Immunoassay
Immunoblot - Size:100 µl
- Pk:100 µl
Specifications
About this item
This mouse Anti-Chondroitin-4-Sulphate is used in ELISA, Radioimmunoassay, Western Blotting techniques and Immunohistochemistry.
- Dilution range: 1:400 to 1:3200 for ELISA
- Dilution range: 1:50 to 1:6400 for RIA
- Dilution range: 1:1000 to 1:4000 for western blot
Type: Primary
Antigen: Chondroitin-4-Sulphate
Clonality: Monoclonal
Clone: BE-123
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1k
Reactivity: Bovine, Dog, Human, Mouse, Rat, Sheep