ɑ-Amino-γ-butyrolactone hydrobromide, off-white powder
Catalog # ICNA0215738905
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- Melting Point:219 - 226°C
- Optical Rotation:~0° (c=20, water)
Specification Test Results
About this item
:
:182.02 g/mol
CAS Number:6305-38-0
Formula:C₄H₇NO₂
MDL Number:MFCD00012723