Human angiotensin II acetate, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Soluble in 0,3 mg/ml, distilled water.
Angiotensin II plays an important role in the regulation of blood pressure. It stimulates the release of aldosterone from the adrenal gland.
Specifications
- CAS No.:68521-88-0
- Species:Human
- Storage conditions:−20 °C
- Form:Powder
- Protein synonyms:H-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-OH|Hypotensin II
- Protein/peptide name:Angiotensin II Acetate
- Molecular weight:1046 kDa