
Eosin Y (yellowish), red powder C.I. 45380
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:17372-87-1
:
Formula:C₂₀H₆Br₄Na₂O₅
MDL Number:MFCD00005040
:691.86 g/mol
:GHS07
Specification Test Results
- UV Visible Absorbance λ max:515 - 518 nm