Bovine lipoprotein-cholesterol concentrate (from Serum), MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Lipoprotein- cholesterol is a mixture of lipoprotein cholesterol, phospholipids and fatty acids.
- Concentration: Cholesterol concentration: 1000 to 2000 mg/dl
- Filtered through 0,2 µm membrane filter
For use as a serum-free or serum-reduced media nutrient supplement, calibration of cholesterol values, and enzymatic or non-enzymatic cholesterol determinations.
Specifications
- Source:Serum
- Species:Bovine
- Storage conditions:Store at −0 °C
- Form:Frozen Golden-Yellow to Orangish Liquid
- Protein/peptide name:Lipoprotein-cholesterol concentrate
- pH:7,5 - 8,0 (Lit.)