Order Entry
Singapore
ContactUsLinkComponent
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from fish sperm)
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from fish sperm)
Supplier:  MP Biomedicals
Certificates
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from fish sperm)
Supplier:  MP Biomedicals

Specification Test Results

  • Synonyms:
    DNA Sodium Salt; DNA; Desoxiribon; Eucytol
  • Form:
    Cream Colored Powder
  • Storage:
    Store at +4 °C, Desiccate
  • Loss on Drying:
    ≤15%
  • Source:
    Salmon or Herring Sperm

Specification Test Results