



Naphthalene ≥99%, white crystalline powder for liquid scintillation counting
Catalog # ICNA0215172680
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- Melting Point:78 - 83 °C
- Purity:≥99%
Specification Test Results
About this item
:
:128.17 g/mol
UN:1334
ADR:4.1,III
:GHS02
:GHS07
:GHS08
:GHS09
CAS Number:91-20-3
Formula:C₁₀H₈
MDL Number:MFCD00001742